Learn More
Ohaus™ Round Plate Clamp
OHAUS™ Round Plate Clamp provides secure support for holding plates at an elevated position.
Merk: Ohaus™ 30392342
77.90 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
- For use with round plates
- Constructed of aluminum
Specificatie
Plate Clamp | |
Aluminum | |
Clamp, Support, Round Plate, CLR-PLATEAR | |
152 mm |