Learn More
BUCHI FlashPure ID Amino Flash Cartridges
Better peak shapes with polar and basic compounds, ideal for carbohydrates
Brand:  BUCHI 145157333
 View more versions of this product
Product Code. 16300296
  • 289.00€ / Each
Only null left
Estimated Shipment: 18-07-2025
Add to basket
Includes: 1 Piece
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications
Specifications
Silica - Amine
1 Pc.
Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
Reveleris
80 g
200 psig