Learn More
Afora™ Reducing Adapters
44.00€ - 111.00€
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
11926638
|
Afora™
3078/E |
M-24 /H-14 | N/A | ||||||
12094952
|
Afora™
9612 |
8/9-12/13 | N/A | ||||||
11946628
|
Afora™
3076 |
M-45 /H-29 | N/A | ||||||
11956648
|
Afora™
3085/AM |
M-14/H-29 AM | N/A | ||||||
11946648
|
Afora™
3085 |
M-14/H-29 |
60.00€
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
11966658
|
Afora™
3090 |
M-19 /H-29 | N/A | ||||||
11916658
|
Afora™
3088 |
M-14/H-19 | N/A | ||||||
12088644
|
Afora™
V20518 |
H-14/23, M-45/40 | N/A | ||||||
11996658
|
Afora™
3091 |
M-24 /H-29 | N/A | ||||||
11916628
|
Afora™
3074/AM |
M-29 /H-14 |
111.00€
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
11906628
|
Afora™
3074 |
M-29 /H-14 |
71.00€
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
11966638
|
Afora™
3080/AM |
M-29 /H-19 AM | N/A | ||||||
11976628
|
Afora™
3077 |
M-19 /H-14 | N/A | ||||||