Learn More
Medline Scientific™ Reciprocal Shaking and Heating Bath
With precise and constant temperature control, providing ideal sample incubation conditions suitable for DNA, tissue or cell culture applications.
Merk: Medline Scientific™ AAH44062K
This item is not returnable.
View return policy
Beschrijving
- Rapid and accurate shaking by adopting silent and anti-vibration brushless DC motors
- +5° above room temperature to 100°C
- Microprocessor PID control/auto tuning/calibration
- Digital timer for delayed ON/OFF from 1 min to 99 hr 59 min
- Protection against over-temperature
- Digital LED display
- Splash-proof keypad
- Separate control keys for shaker mechanism allowing the unit also to be used as a constant temperature bath
- Minimal shock to the sample thanks to the shaker’s gentle start feature
- Multiple LED displays for setting various values
- Isolated design of agitator, heater and sensor by a baffle plate to minimize the turbulence in the bath fluid and also to protect users and samples against direct contact
- Removable shaking carriage designed for external assembly avoiding direct contact with the bath fluid
- Minimization of the liquid loss and dripping of the bath liquid into the samples thanks to the included gable cover
- Included SUS 304 spring wire rack with double-two-tier providing various ways of glassware settings
Specificatie
Reciprocating motion; 20 to 180 rpm | |
Ambient +5° to 100°C | |
DNA, tissue, cell culture applications | |
BS-06 | |
440 x 430 x 355 mm | |
230 V |
17 L | |
LED | |
Stainless steel gable cover, spring wire rack | |
±0.2° | |
26 kg | |
50 Hz |