Learn More
RBM22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 22103-1-AP-150UL
This item is not returnable.
View return policy
Beschrijving
RanGAP1, is a homodimeric 65-kD polypeptide that specifically induces the GTPase activity of RAN, but not of RAS by over 1, 000-fold. RanGAP1 is the immediate antagonist of RCC1, a regulator molecule that keeps RAN in the active, GTP-bound state. The RANGAP1 gene encodes a 587-amino acid polypeptide. The sequence is unrelated to that of GTPase activators for other RAS-related proteins, but is 88% identical to Fug1, the murine homolog of yeast Rna1p. RanGAP1 and RCC1 control RAN-dependent transport between the nucleus and cytoplasm. RanGAP1 is a key regulator of the RAN GTP/GDP cycle.Specificatie
RBM22 | |
Polyclonal | |
Unconjugated | |
RBM22 | |
fSAP47, Pre mRNA splicing factor RBM22, RBM22, RNA binding motif protein 22, ZC3H16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
307410, 55696, 66810 | |
-20°C | |
Liquid |
Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4V7D7, Q8BHS3, Q9NW64 | |
RBM22 | |
RBM22 Fusion Protein Ag17315 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |