Learn More
RAD23A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk:  Proteintech 11364-1-AP-150UL
 Bekijk meerdere types van dit product via de
Produktcode. 16898383
  • 410.00 / 150µL
Beschrijving
Beschrijving
This antibody is a rabbit polyclonal antibody raised against full length RAD23A of human origin.
The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in nucleotide excision repair (NER). This protein was shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, as well as with ubiquitin protein ligase E6AP, and thus suggests that this protein may be involved in the ubiquitin mediated proteolytic pathway in cells.
Specificatie
Specificatie
RAD23A
Polyclonal
Unconjugated
RAD23A
HHR23A, HR23A, RAD23A
Rabbit
Antigen Affinity Chromatography
RUO
19358, 361381, 5886
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P54725, P54726
Rad23a
RAD23A Fusion Protein Ag1907
150 μL