Learn More
PRKACA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk:  Proteintech 24503-1-AP-20UL
 Bekijk meerdere types van dit product via de
Produktcode. 16882975
  • 145.00 /
Explore more special offers
Beschrijving
Beschrijving
The PRKACA antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PRKACA. This antibody recognizes human, mouse, rat antigen. The PRKACA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specificatie
Specificatie
PRKACA
Polyclonal
Unconjugated
20 μL
5566
IgG
Western Blot, Immunohistochemistry, ELISA
333 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen