Learn More
Masterflex™ Tee-Connector Compression Fitting, polypropylene
Connect rigid tubing systems with these tee connectors. Polypropylene Tee-Connector Compression Fittings form a tight seal and withstand higher pressures than barbed fittings.
34.85CHF - 170.00CHF
Specifications
Material | Polypropylene |
---|---|
Min. Operating Temperature (Metric) | 0°C |
Packaging Quantity | 10 Pack |
Maximum Pressure | 220 psi |
Max. Operating Temperature (Metric) | 100°C |
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
11781889
|
Masterflex™
06382-40 |
1/2 in. tubing O.D. |
170.00CHF
Pack of 10 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
11761759
|
Masterflex™
06382-10 |
1/4 in. tubing O.D. | N/A | ||||||
11751759
|
Masterflex™
06382-05 |
1/8 in. tubing O.D. | N/A | ||||||
11791879
|
Masterflex™
06382-30 |
3/8 in. tubing O.D. | N/A | ||||||
Description
Assemble fittings to tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.- Excellent chemical resistance
- Rated to 220 psi maximum at 25 °C
- Usable from 0 to 100 °C
- Sterilization: Ethylene Oxide (ETO)
Specifications
Polypropylene | |
10 Pack | |
100°C |
0°C | |
220 psi | |
Compression Fitting |