Learn More
PKM1 Rabbit anti-Human, Mouse, Rat, Biotin, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech BIOTIN-15821-100UL
This item is not returnable.
View return policy
Beschrijving
This antibody is conjugated with Biotin and it's specific to PKM1 isoform.
PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.Specificatie
PKM1 | |
Polyclonal | |
Biotin | |
PKM | |
CTHBP, OIP 3, OIP3, Opa interacting protein 3, p58, PK2, PK3, PKM, PKM1, PKM2, PKM2 pyruvate kinase, muscle, Pyruvate kinase 2/3, Pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, TCB, THBP1, Tumor M2 PK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18746, 25630, 5315 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.38 mg/mL | |
PBS with 50% glycerol and no preservative; pH 7.3 | |
P11980, P14618, P52480 | |
Pkm | |
Peptide | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |