Learn More
PHOX2B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk:  Proteintech 25276-1-AP-150UL
 Bekijk meerdere types van dit product via de
Produktcode. 16853855
  • 410.00 /
Beschrijving
Beschrijving
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.
Specificatie
Specificatie
PHOX2B
Polyclonal
Unconjugated
PHOX2B
NBPhox, Neuroblastoma Phox, paired like homeobox 2b, PHOX2B, PHOX2B homeodomain protein, PMX2B
Rabbit
Antigen Affinity Chromatography
RUO
18935, 8929
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35690, Q99453
Phox2b
PHOX2B Fusion Protein Ag13196
150 μL