Learn More
pH 7.41 Buffer Solution at 25°C According to European Pharmacopoeia (EP), Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
Description | Accuracy ± 0.01 pH units |
pH | 7.41 ±0.01 |
Physical Form | Liquid |
Quantity | 500 mL |
Packaging | HDPE Bottle GL45 |