Learn More
Bode Chemie™ Peha-Soft™ Powder-Free Glove
Peha-Soft™ powder-free examination glove made of latex.
14.00
Specificatie
Certificeringen/conformiteit | Approved for Contact with Food |
---|---|
Kleur | White |
Voor eenmalig gebruik | Single-use |
Latex Free | No |
Materiaal | Latex |
Beschrijving
- Disposable examination gloves made of soft latex, powder-free
- Easy to put on thanks to the inner coating
- Textured finger area for optimal grip
- Highly elastic, tear-resistant and safe
- Excellent fit and excellent tactile sensitivity
- To wear left and right.
Specificatie
Approved for Contact with Food | |
Single-use | |
Latex | |
Examination Gloves |
White | |
No | |
Powder Free |