Learn More
PCMT1 Mouse anti-Human, CoraLite 488, Clone: 3C8G8, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech CL488-60172-100UL
This item is not returnable.
View return policy
Beschrijving
The PCMT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PCMT1. This antibody recognizes human antigen. The PCMT1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specificatie
PCMT1 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence | |
3C8G8 | |
CoraLite 488 | |
100 μL | |
5110 | |
IgG1 |