Learn More
Thermo Scientific™ A2 and A5 Large Gel System Replacement End Gates
Use these gasketed UVT gel trays, replacement gaskets and end gates with Thermo Scientific™ A2 and A5 Large Gel Systems.
Merk: Thermo Scientific™ A2-EG
This item is not returnable.
View return policy
Beschrijving
Use Thermo Scientific™ A2 and A5 Large Gel System Replacement Parts with Owl A2 and A5 Large Gel Systems.
- UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
- Comb slots are positioned to allow 2, 3 or 4 sample sets to run equal lengths on the same gel
Specificatie
Owl A2 and A5 Large Gel Electrophoresis System | |
End Gates | |
1 Pair |