Learn More
3M™ Oil Absorbent Sheets
3M™ Oil Absorbent Sheets ideal for absorbing small oil leaks and quickly cleaning equipment.
Merk: 3M™ 7000088323
This item is not returnable.
View return policy
Beschrijving
- Hydrophobic
- Repel water
- Ideal for use in areas where water may be present
Specificatie
100/Cs. | |
390 x 530 mm |