Learn More
NAT10 Rabbit, anti-Human, CoraLite Plus 488, Clone: 5B9, Proteintech
Rabbit Recombinant Monoclonal Antibody
Merk: Proteintech CL488-82585-100UL
This item is not returnable.
View return policy
Beschrijving
The NAT10 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human NAT10. This antibody recognizes human antigen. The NAT10 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specificatie
NAT10 | |
5B9 | |
CoraLite Plus 488 | |
Rabbit | |
Primary | |
Human | |
150 kDa | |
IgG |
Immunofluorescence | |
1000 μg/mL | |
ALP, DKFZp434C116, FLJ10774, FLJ12179, FLJ23850, KIAA1709, N acetyltransferase 10, NAT10 | |
100 μL | |
55226 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |