Learn More
MVK Mouse anti-Human, Unconjugated, Clone: 2F1D3, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech 67820-1-IG-20UL
This item is not returnable.
View return policy
Beschrijving
The MVK antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human MVK. This antibody recognizes human antigen. The MVK antibody has been validated for the following applications: WB, ELISA analysis.
Specificatie
MVK | |
Monoclonal | |
Unconjugated | |
20 μL | |
4598 | |
IgG2b |
Western Blot, ELISA | |
2F1D3 | |
Mouse | |
Primary | |
Human |