Learn More
MED20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 17598-1-AP-150UL
This item is not returnable.
View return policy
Beschrijving
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.Specificatie
MED20 | |
Polyclonal | |
Unconjugated | |
MED20 | |
hTRFP, MED20, mediator complex subunit 20, PRO0213, TRF proximal protein homolog, TRFP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
316209, 56771, 9477 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5XIE9, Q9H944, Q9R0X0 | |
Med20 | |
MED20 Fusion Protein Ag11598 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |