missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKSP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 11937-1-AP-20UL
This item is not returnable.
View return policy
Beschrijving
This gene encodes a scaffold protein that functions in the extracellular signal-regulated kinase (ERK) cascade. The protein is localized to late endosomes by the mitogen-activated protein-binding protein-interacting protein, and binds specifically to MAP kinase kinase MAP2K1/MEK1, MAP kinase MAPK3/ERK1, and MAP kinase MAPK1/ERK2. Studies of the orthologous gene in mouse indicate that it regulates late endosomal traffic and cell proliferation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. A pseudogene of this gene is located on the long arm of chromosome 13.Specificatie
MAPKSP1 | |
Polyclonal | |
Unconjugated | |
LAMTOR3 | |
LAMTOR3, MAP2K1IP1, MAPBP, MAPK scaffold protein 1, MAPKSP1, MEK binding partner 1, MP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362045, 56692, 8649 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88653, Q5U204, Q9UHA4 | |
Lamtor3 | |
MAPKSP1 Fusion Protein Ag2534 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction