missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAG Mouse anti-Human, Mouse, Rat, Clone: 3H9G3, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech 66709-1-IG-20UL
This item is not returnable.
View return policy
Beschrijving
The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.Specificatie
MAG | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P07722, P20916, P20917 | |
Mag | |
MAG Fusion Protein Ag6065 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG1 |
Western Blot | |
3H9G3 | |
Unconjugated | |
MAG | |
GMA, MAG, myelin associated glycoprotein, S MAG, SIGLEC 4A, SIGLEC4A | |
Mouse | |
Protein G | |
RUO | |
17136, 29409, 4099 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction