Learn More
DWK Life Sciences Kimble™ Kontes™ Extraction Manifold Replacement Components
For extraction manifold.
Merk:  DWK Life Sciences 971101-1047
 Bekijk meerdere types van dit product via de
Produktcode. 10704017
  • 245.00 / 1 stuk
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • For use with Kontes SPE Manifold Sets (971000-1047, -3047, or -6047)
  • Kontes SPE Glassware and Manifolds are specially designed as support systems for 47mm and 90mm dia. solid phase extraction disks to give you the highest possible recoveries of extracted analytes from your drinking water, surface water and waste water samples
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
1000 mL
Glass Funnel (1000 mL)
Glass
4.7 cm
Solid Phase Extraction Manifold
47 mm (1000 mL) Funnel
1.85 in.
47 mm