Learn More
DWK Life Sciences™ Kimble™ Sample Vials with Black Phenolic Caps and Polyvinyl-faced Pulp Liners
Kimble™ Screw thread N-51A Borosilicate Glass Sample Vials made of low extractable Kimax N-51A glass.
417.00 - 623.00
Specificatie
Kleur van sluiting | Black |
---|---|
Kleur | Clear |
Materiaal van sluiting | Phenolic |
Produktcode | Merk | Volume (metrisch) | Afmetingen van sluiting | Diameter (metrisch) buiten | Prijs | Hoeveelheid en beschikbaarheid | |||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Volume (metrisch) | Afmetingen van sluiting | Diameter (metrisch) buiten | Prijs | Hoeveelheid en beschikbaarheid | |||
16415089
|
DWK Life Sciences™
60910C-6 |
24 mL | 20-400 | 23 mm |
623.00
|
|
|||
15960471
|
DWK Life Sciences™
60910C-8 |
30 mL | 22-400 | 25 mm |
417.00
|
|
|||
Beschrijving
Kimble™ Screw-Thread N-51A Borosilicate Glass Sample Vials made of low extractable Kimax N-51A glass.
- Vials with phenolic caps and polyvinyl-faced liners
- Borosilicate glass sample vials with caps
- Caps are not attached
- Clear screw thread sample vial
- Closed-top black phenolic closure with polyvinyl-faced pulp liner (not attached)
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Compliance
Kimax N-51A glass conforms to ASTM Type 1, Class B requirements.
Specificatie
Black | |
Clear | |
Phenolic |