Learn More
Iron solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
Product Code. 12947353 Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
Unit Size:
100mL
50mL
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
Specifications
Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix HNO3
Quantity 50 mL
Chemical Name or Material Iron