Learn More
ILK Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk:  Proteintech 12955-1-AP-20UL
 Bekijk meerdere types van dit product via de
Produktcode. 16870934
  • 145.00 /
Explore more special offers
Beschrijving
Beschrijving
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. ILK is a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction.
Specificatie
Specificatie
ILK
Polyclonal
Unconjugated
ILK
ILK, ILK 1, ILK 2, ILK1, ILK2, P59, p59ILK
Rabbit
Antigen Affinity Chromatography
RUO
16202, 170922, 3611, 393543
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O55222, Q13418, Q99J82
Ilk
ILK Fusion Protein Ag3570
20 μL
Primary
Human, Rat, Mouse, Zebrafish
Antibody
IgG
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen