missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILK Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 12955-1-AP-150UL
This item is not returnable.
View return policy
Beschrijving
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. ILK is a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction.Specificatie
ILK | |
Polyclonal | |
Unconjugated | |
ILK | |
ILK, ILK 1, ILK 2, ILK1, ILK2, P59, p59ILK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16202, 170922, 3611, 393543 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O55222, Q13418, Q99J82 | |
Ilk | |
ILK Fusion Protein Ag3570 | |
150 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction