Learn More
Proteintech IHCeasy LDB1 Ready-To-Use IHC Kit
KHC1808 is a ready-to-use IHC kit for staining of LDB1.
Brand:  Proteintech KHC1808-50T
Product Code. 18099325
  • 389.00€ / 50 tests
Explore more special offers
Description
Description
KHC1808 is a ready-to-use IHC kit for staining of LDB1. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
50 Tests
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers