missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-757 Safety Gloves

The HyFlex™ 11-757 Safety Gloves are ultra-thin breathable gloves featuring high level cut protection and touchscreen compatibility.

346.38

Specificatie

Certificeringen/conformiteit ANSI ABR 4, ANSI CUT A7
Certifications/Compliance (Europe) EN 388:3X31F, EN 420:2003 + A1:2009
Coating Material Polyurethane
Kleur Black
Hand Specific
Meer specificaties weergeven

Producten 1
Produktcode Merk Maat Prijs Hoeveelheid en beschikbaarheid  
Produktcode Merk Maat Prijs Hoeveelheid en beschikbaarheid  
30259529
Bekijk documenten
Ansell
11757090
9
346.38
Only null left
Toevoegen aan winkelmandje
 
Beschrijving

Beschrijving

  • Extreme cut protection featuring INTERCEPT™ Technology while maintaining enhanced flexibility and tactility
  • Ultra-lightweight and ultra-thin breathable gloves featuring touchscreen compatibility minimize the need to change or remove gloves
  • High dexterity combined with unmatched comfort and precision while achieving excellent abrasion resistance
  • Tailored fit at the base of the pinky finger better matches natural hand contours
  • Dermatologically approved by Dermatest™
Specificatie

Specificatie

ANSI ABR 4, ANSI CUT A7
Polyurethane
Specific
Nylon, Stainless Steel Fiber, Polyester, Tungsten
Knit
8.7 to 10.8 in.
EN 388:3X31F, EN 420:2003 + A1:2009
Black
Yes
Cut-resistant Gloves
11-757
22.2 to 27.4 cm
Video's
SDS
Documentatie

Documentatie

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.