Learn More
Krüss Optronic™ HR10 Model Manual Handheld Refractometer
Brand: Krüss Optronic™ HR10
This item is not returnable.
View return policy
Specifications
Handheld Manual Refractometer | |
HR10 | |
Sugar concentration in fruit juices, soft drinks, vegetables, foods and cooling lubricants |
0 to 10 Brix | |
0.1% Brix | |
Supplied in a case, complete with conversion tables |