Learn More
Thermo Scientific Chemicals Guanosine 5'-triphosphate, disodium salt hydrate, 90%, for biochemistry
Guanosine 5'-triphosphate, disodium salt hydrate, 90%, C10H14N5Na2O14P3.xH2O, CAS Number-56001-37-7
53.70 - 584.00
Chemische identificatiekenmerken
CAS | 56001-37-7 |
---|---|
Moleculaire formule | C10H14N5Na2O14P3 |
Molecular Weight (g/mol) | 567.14 |
MDL-nummer | MFCD03410297 |
InChI Key | FIZIYLKEXVIRHJ-KHRSEZDTNA-L |
Synoniem | guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate |
PubChem CID | 131676145 |
IUPAC Name | disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate |
SMILES | [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1 |
Produktcode | Merk | Aantal | Verpakking | Prijs | Hoeveelheid en beschikbaarheid | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Aantal | Verpakking | Prijs | Hoeveelheid en beschikbaarheid | ||||
10184750
|
Thermo Scientific Chemicals
226250500 |
50 mg | Glass bottle |
53.70
50 mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10012880
|
Thermo Scientific Chemicals
226252500 |
250 mg | Glass bottle |
169.00
250 mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10732461
|
Thermo Scientific Chemicals
226250010 |
1 g | Glass bottle |
584.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschrijving
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistryChemische identificatiekenmerken
56001-37-7 | |
567.14 | |
FIZIYLKEXVIRHJ-KHRSEZDTNA-L | |
131676145 | |
[Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1 |
C10H14N5Na2O14P3 | |
MFCD03410297 | |
guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate | |
disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate |
Specificatie
Guanosine 5'-triphosphate, disodium salt hydrate | |
1.12 to 1.18 (A250/A260), 0.63 to 0.69 (A280/A260) | |
White | |
88% min. (HPLC) | |
MFCD03410297 | |
Solubility in water: soluble | |
FIZIYLKEXVIRHJ-KHRSEZDTNA-L | |
disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate | |
131676145 | |
90% | |
Biochemistry |
56001-37-7 | |
180°C | |
Authentic | |
C10H14N5Na2O14P3 | |
guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate | |
88% min. (lambda max. 252nm,pH 7) | |
[Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1 | |
567.14 | |
567.14 | |
4% max. (typically) Guanosine diphosphate (HPLC) | |
Crystalline Powder |
Veiligheid en behandeling
missing translation for 'einecsNumber' : 259-940-7
Productidentificatie
- Guanosine 5'-triphosphate disodium salt hydrate