Learn More
Afora™ Graduated Cylinders
81.00€ - 298.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12047950
|
Afora™
V64877 |
250 mL | N/A | ||||||
12092430
|
Afora™
V62260 |
100 mL |
81.00€
Each |
Estimated Shipment: 27-10-2025
Log in to see stock. |
|||||
12037033
|
Afora™
V53827 |
1000 mL |
185.00€
Each |
Estimated Shipment: 01-10-2025
Log in to see stock. |
|||||
Specifications
Glass | |
Yes | |
Graduated Cylinder |