Learn More
Afora™ Funnel for General Use
Prevent spillage with the Afora™ Funnel for General Use. A funnel is a pipe with a wide (often conical) mouth and a narrow stem.
52.00€ - 67.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12018892
|
Afora™
6395/10 |
10 mL |
67.00€
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||||
12038892
|
Afora™
6395/3 |
3 mL |
52.00€
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
12058892
|
Afora™
6395/6 |
6 mL |
56.00€
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
Description
- Glass
Specifications
Glass | |
Funnel |