Learn More
Testo™ Frozen food probe
Merk: Testo™ 0603 3292
157.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specificatie
Frozen food probe | |
Thermocouple type T | |
Measuring the core temperature of frozen food | |
1.7 m plug-in cable |