Learn More
Poulten & Graf GmbH FORTUNA™ Precision Gas Syringe
Poulten & Graf GmbH FORTUNA™ Precision Gas Syringe are used to measure gas volume.
Merk: Poulten & Graf GmbH 2 01251
This item is not returnable.
View return policy
Beschrijving
- Clear neutral glass
- Calibrated cylinders
- Acid and alkali resistant amber stain diffused graduation
- Amber graduation
Specificatie
Autoclavable | |
Clear | |
FORTUNA Precision Gas Syringe with straight stopcock | |
Amber | |
100 mL | |
Non-sterile |
Glass (Tip), Neutral Glass, Neutral Glass (Barrel), Neutral Glass (Plunger) | |
Straight Stopcock | |
Reusable | |
1 | |
Yes |