Learn More
SCAT Flat Canisters
Click to view available options
Capacity (Metric):
8 L
20 L
Closure Size:
63B
S70/71
Dimensions (L x W x H):
283 x 376 x 292 mm
375 x 305 x 165 mm
Description
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in laboratories
- Available with B63 or S70/71 threads and can be adapted to many other thread sizes by means of an adapter
- Available either as compact 8 L canister or as more voluminous 20 L canister
SCAT flat canisters are an ideal way of not having to dispense with safe storage even when space is limited, as they can be integrated into laboratory furniture to save space. SCAT flat pack canisters are also optimally available with UN-Y approval for the storage of hazardous goods or with an integrated drain cock for safe emptying.
Specifications
Specifications
Capacity (English) | 2 gal. |
Capacity (Metric) | 8 L |
Closure Size | 63B |
Color | White (Translucent) |
Description | Flat can with drain valve, 8 L, B63, PP, dimensions: (W x H x D): 375 x 165 x 305 mm, autoclavable |
Dimensions (L x W x H) | 375 x 305 x 165 mm |
Includes | with drain valve, autoclavable |
Material | Polypropylene |
Mouth Style | B63 Thread |
Product Type | Canister |