Learn More
Grant Instruments™ Flask Tray for Grant Shaking Water Baths
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruments™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
Merk: Grant Instruments™ TF12
313.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Beschrijving
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruments™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
- Accommodates flasks from 25 to 1000mL
- For use with the followng Shaking Water Bath models:
- LSB12
- LSB18
- OLS26
- Compatible with SH plate holders or SR racks
- TF26 Test tube tray also compatible with SR racks
- Can be used alone to accommodate bags and miscellaneous vessels.
Specificatie
25 to 1000 mL | |
LSB12 | |
Flask Tray |