Learn More
BUCHI FlashPure ID C18-WP Chromatography Cartridges
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
157.00€ - 1150.00€
Specifications
Material | Polypropylene |
---|---|
Type | C18 Wide Pore |
Includes | 1 Pc. |
Resolution | 16 to 24 μm |
Column Type | C18-WP |
Product Code | Brand | Size (Metric) Tube Rack | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) Tube Rack | Includes | Price | Quantity & Availability | ||||
16370256
|
BUCHI
140000153 |
120 g | 1 Pc. |
471.00€
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
16380256
|
BUCHI
140000154 |
220 g | 1 Pc. | N/A | |||||
16310146
|
BUCHI
140000019 |
24 g | 1 Pc. |
157.00€
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
16390256
|
BUCHI
140000155 |
330 g | 1 Pc. |
1150.00€
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
16360256
|
BUCHI
140000152 |
80 g | 1 Pc. |
305.00€
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
Description
- High resolution of separation due to high-quality silica
- Increased fraction purity due to a better separation
- Faster evaporation of smaller fraction volumes
- Reproducible performance from batch to batch
- Increased sample throughput due to high sample loading capacity
- Solvent saving: required purification yield is achieved faster
Specifications
Polypropylene | |
1 Pc. | |
C18-WP | |
Reveleris Flash System |
C18 Wide Pore | |
16 to 24 μm | |
Wide Pore Media for Large Molecules, Ideal for Peptides, Protiens and Lipids |