Learn More
Witeg™ Filtration Flask with Plastic Hose Connection and Tubulature
Collect filtrate in these highly durable flasks. Witeg™ Filtration Flask with Plastic Hose Connection and Tubulature is made of highly resistant Duran™ with a Levasint™ safety coating.
Merk:  Witeg™ 5 071 900
Produktcode. 10041732
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »
Explore more special offers
Beschrijving
Beschrijving
  • Hose connection and tubulature.
  • Duran™ construction.
  • Levasint™ safety coating.
Specificatie
Specificatie
10,000 mL
Plastic hose connection and tubulature
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen