Learn More
Endothelin 3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 27871-1-AP-20UL
This item is not returnable.
View return policy
Beschrijving
The Endothelin 3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Endothelin 3. This antibody recognizes human, mouse, rat antigen. The Endothelin 3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specificatie
Endothelin 3 | |
550 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat | |
150 kDa | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
EDN3, endothelin 3, ET 3, ET3, PPET3, Preproendothelin 3 | |
20 μL | |
1908 | |
Store at -20°C. Stable for one year after shipment. | |
Liquid |