Learn More
Thermo Scientific™ Dionex™ AS-DV Autosampler PolyVials and Caps
Employ convenient packs of 0.5mL or 5.0mL sample vials, plain or filter caps, and vial accessories for the Thermo Scientific™ Dionex™ AS-DV Autosampler.
Brand:  Thermo Scientific™ 038009
Encompass
 View more versions of this product
Product Code. 12819772
  • 216.00 € / Pack of 250
Description
Available in 0.5 mL and 5.0 mL sizes, Thermo Scientific™ Dionex™ AS-DV Autosampler PolyVials are supplied in packs of 250, with and without plain or filter caps. Choose from compatible vial holders, adapters and a vial cap removal tool to complete your setup for the AS-DV Autosampler.
Ordering Information:
Supplied in packs of 250, with and without caps
Specifications
Specifications
Filter Caps
5 mL AS-DV PolyVials
Each 250
Filter caps only
5 mL
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.