missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 16275-1-AP-20UL
This item is not returnable.
View return policy
Beschrijving
This gene encodes a member of the short-chain dehydrogenases/reductases (SDR) family. The encoded enzyme contains a conserved catalytic domain and likely functions as an oxidoreductase. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specificatie
DHRS1 | |
Polyclonal | |
Unconjugated | |
DHRS1 | |
DHRS1, SDR19C1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115817, 290234, 52585 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96LJ7, Q99L04 | |
Dhrs1 | |
DHRS1 Fusion Protein Ag9351 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction