Learn More
DGCR8 N-terminal Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 25835-1-AP-20UL
This item is not returnable.
View return policy
Beschrijving
This gene encodes a subunit of the microprocessor complex which mediates the biogenesis of microRNAs from the primary microRNA transcript. The encoded protein is a double-stranded RNA binding protein that functions as the non-catalytic subunit of the microprocessor complex. This protein is required for binding the double-stranded RNA substrate and facilitates cleavage of the RNA by the ribonuclease III protein, Drosha. Alternate splicing results in multiple transcript variants.Specificatie
DGCR8 N-terminal | |
Polyclonal | |
Unconjugated | |
DGCR8 | |
C22orf12, DGCR8, DGCRK6, Gy1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54487 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WYQ5 | |
DGCR8 | |
DGCR8 N-terminal Fusion Protein Ag22988 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |