Learn More
DEPDC6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 21312-1-AP-20UL
This item is not returnable.
View return policy
Beschrijving
Negative regulator of the mTORC1 and mTORC2 signaling pathways. Inhibits the kinase activity of both complexes.Specificatie
DEPDC6 | |
Polyclonal | |
Unconjugated | |
DEPTOR | |
DEP domain containing 6, DEP.6, DEPDC6, Deptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64798 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TB45 | |
DEPTOR | |
DEPDC6 Fusion Protein Ag15959 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |