Learn More
Thermo Scientific™ Culti-Loops™ Naganishia albida var. albidus ATCC™ 10666™
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
ATCC® | 10666TM* |
Description | Naganishia albida var. albidus |
Product Type | Culti-Loops Quality Control Organism |
Target Organism Class | Fungi |
Test Type | Qualitative QC |
Quantity | 5 Loops/Pack |
Strain Designations | NNRL, Y-1400 [CBS 142, CCRC 21672, CCY 17-14-1, IFO 0378, IFO 0610, JCM 2334, MUCL 30400, NCYC 445, VKM Y-2223] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.