Learn More
CPT1A Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk:  Proteintech CL555-15184-100UL
Produktcode. 17561636
  • 450.00 /
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Explore more special offers
Beschrijving
Beschrijving
The CPT1A antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CPT1A. This antibody recognizes human, mouse, rat antigen. The CPT1A antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™555.
Specificatie
Specificatie
CPT1A
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Flow Cytometry (Intracellular Staining)
CoraLite 555
100 μL
1374
IgG
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen