missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COTL1 Mouse anti-Human, Mouse, Porcine, Clone: 4A3D9, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech 60237-1-IG-150UL
This item is not returnable.
View return policy
Beschrijving
This gene encodes one of the numerous actin-binding proteins which regulate the actin cytoskeleton. This protein binds F-actin, and also interacts with 5-lipoxygenase, which is the first committed enzyme in leukotriene biosynthesis. Although this gene has been reported to map to chromosome 17 in the Smith-Magenis syndrome region, the best alignments for this gene are to chromosome 16. The Smith-Magenis syndrome region is the site of two related pseudogenes.Specificatie
COTL1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14019, Q9CQI6 | |
Cotl1 | |
COTL1 Fusion Protein Ag1230 | |
150 μL | |
Primary | |
Human, Mouse, Pig | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
4A3D9 | |
Unconjugated | |
COTL1 | |
CLP, Coactosin like protein, COTL1 | |
Mouse | |
Protein A | |
RUO | |
100621773, 23406, 72042 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction