missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 11457-1-AP-150UL
This item is not returnable.
View return policy
Beschrijving
The product of this gene is an epsilon subunit of coatomer protein complex. Coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles. It is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. Coatomer complex consists of at least the alpha, beta, beta′, gamma, delta, epsilon and zeta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.Specificatie
COPE | |
Polyclonal | |
Unconjugated | |
COPE | |
Coatomer subunit epsilon, COPE, Epsilon coat protein, epsilon COP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11316 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14579 | |
COPE | |
COPE Fusion Protein Ag2024 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction