Learn More
Memmert™ Clip for 200 mL Flask
Clip for 200 mL flask (14 flasks max.)
Merk: Memmert™ WTB35B02693
27.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
- Accessory
- Max. flasks see sketch on demand
Specificatie
WTB35 | |
Clip for 200 mL flask |