Learn More
BRAND™ Class A Amber Borosilicate Volumetric Flask with Glass Stopper
Amber volumetric flask with glass stopper and USP certificate.
85.55 - 136.00
Specificatie
Kalibratie | To Contain |
---|---|
Materiaal | Borosilicate Glass (3.3) |
Autoclaveerbaar | Autoclavable |
Voor eenmalig gebruik | Reusable |
Graduated | Yes |
Produktcode | Merk | Inhoud (metrisch) | Certificeringen/conformiteit | Prijs | Hoeveelheid en beschikbaarheid | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Inhoud (metrisch) | Certificeringen/conformiteit | Prijs | Hoeveelheid en beschikbaarheid | ||||
16357997
|
BRAND™
237464 |
25 mL | USP Single Batch |
90.12
2 stuks |
|
||||
16367997
|
BRAND™
237465 |
50 mL | USP Single Batch |
85.55
Pack of 2 |
|
||||
16377997
|
BRAND™
237467 |
100 mL | USP Single Batch |
86.40
Pack of 2 |
|
||||
16387997
|
BRAND™
237469 |
250 mL | USP Single Batch |
114.00
2 stuks |
|
||||
15131216
|
BRAND™
957469 |
250 mL | USP Single |
87.25
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15177316
|
BRAND™
957471 |
1 L | USP Single |
136.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschrijving
- Borosilicate 3.3
- To DIN EN ISO 1042
- Calibrated to contain (TC, In)
- Includes USP certificate, single, single batch, or batch delivery
Specificatie
To Contain | |
Autoclavable | |
Yes | |
Non-sterile |
Borosilicate Glass (3.3) | |
Reusable | |
Narrow | |
Flat |