Learn More
SPEX Certiprep Clamp for ShaQer. 6 x 50ml or 6 x 2ml tubes
Merk:  SPEX Certiprep 1595
Clamp for ShaQer. 6 x 50ml or 6 x 2ml tubes
Overige gegevens : Weight : 3.00000kg
Produktcode. 15340472
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »