Learn More
CD8a Mouse anti-Human, CoraLite 488, Clone: Hit8a, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech CL488-65113-100TESTS
This item is not returnable.
View return policy
Beschrijving
The CD8a antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8a. This antibody recognizes human antigen. The CD8a antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™488.
Specificatie
CD8a | |
Monoclonal | |
5 μL/Test | |
Mouse | |
Primary | |
Human |
Immunofluorescence, Flow Cytometry | |
Hit8a | |
CoraLite 488 | |
100 Tests | |
925 | |
IgG1 |