Learn More
CD8a Mouse anti-Human, CoraLite 488, Clone: Hit8a, Proteintech
Mouse Monoclonal Antibody
Merk:  Proteintech CL488-65113-100TESTS
Produktcode. 16810666
  • 135.00 /
Explore more special offers
Beschrijving
Beschrijving
The CD8a antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8a. This antibody recognizes human antigen. The CD8a antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™488.
Specificatie
Specificatie
CD8a
Monoclonal
5 μL/Test
Mouse
Primary
Human
Immunofluorescence, Flow Cytometry
Hit8a
CoraLite 488
100 Tests
925
IgG1
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen